Tide Fluor™ 1 succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF1 SE
2244
AATB2244EA
307
EUR
AATB2244
Tide Fluor™ 1 succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 1 succinimidyl ester
TF1 is designed to be a superior fluorophore alternative to EDANS.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
330,29 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...