Tide Fluor™ 2 acid, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF2 acid
2245
AATB2245EA
307
EUR
AATB2245
Tide Fluor™ 2 acid, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2 acid
Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
487,51 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...