Tide Fluor™ 2 amine, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF2 amine

2246
AATB2246EA 307 EUR
AATB2246
Tide Fluor™ 2 amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2 amine

Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 757,63 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7136ef011b9ced237e43307d45d21e73037d620f&vid=fd480084e967b855bc1c3ddbb968bb3768a3224d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR