Tide Fluor™ 2 maleimide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF2 maleimide

2247
AATB2247EA 307 EUR
AATB2247
Tide Fluor™ 2 maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2 maleimide

Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 680,71 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1d791cdeabb97475c22f69c8aa708d777a16b94&vid=6e85ce31240faec2de8c36a4ab93d88cace67150&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR