Tide Fluor™ 2, succinimidyl ester, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF2 SE

2248
AATB2248EA 307 EUR
AATB2248
Tide Fluor™ 2, succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2, succinimidyl ester

Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 584,58 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3bafafd6c9298ebabca1a195817c98b9b754ebc&vid=199e39c2fccdc1683056d838a6435d21b7f66680&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR