Tide Fluor™ 2 azide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF2 azide

2252
AATB2252EA 307 EUR
AATB2252
Tide Fluor™ 2 azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2 azide

Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 783,62 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a3b0fc8d8540efb886024b0d8727655d774fc32&vid=bb09cdfc76e5b711c2b2cce3acf2b16b4ed97be1&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR