Tide Fluor™ 2 alkyne, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT Bioquest
Achtung

2253
AATB2253EA 307 EUR
AATB2253
Tide Fluor™ 2 alkyne, Tide Fluor™ fluoreszierende Probe

Tide Fluor™ 2 (TF2) family has the spectral properties similar to those of fluoresceins and Alexa Fluor® 488 (Alexa Fluor® is the trademark of Invitrogen).


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa171184d0193c824b1472c0104df25bf504d337&vid=fc00850bd9d0a1e6d2ec44334ad1d432738d3419&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR