Tide Fluor™ 3 azide, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF3 azide
2254
AATB2254EA
307
EUR
AATB2254
Tide Fluor™ 3 azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3 azide
Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
526,59 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...