Cell Explorer™ Live Cell Labelling Kit *Green Fluorescence with 405 nm Excitation*
Lieferant: AAT BIOQUEST
22615
AATB22615EA 307 EUR
AATB22615
Cell Explorer™ Live Cell Labelling Kit *Green Fluorescence with 405 nm Excitation*
Tests Zelltests
The Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
  • Contains all necessary components
  • Multiple research applications
  • Large Stokes shift
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Bitte beachten For research use only.