Cell Explorer™ Live Cell Labelling Kit *Green Fluorescence with 405 nm Excitation*
Lieferant: AAT BIOQUEST
22615
AATB22615EA
307
EUR
AATB22615
Cell Explorer™ Live Cell Labelling Kit *Green Fluorescence with 405 nm Excitation*
Tests
Zelltests
The Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- Contains all necessary components
- Multiple research applications
- Large Stokes shift
- Bright and stable signal
- Suitable for all major fluorescence platforms
Bitte beachten For research use only.