LysoBrite™ Blue
Lieferant: AAT BIOQUEST
22642
AATB22642EA
230
EUR
AATB22642
LysoBrite™ Blue
Tests
Zelltests
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Lysosome-Specific Dye
- Long-Term Use
- Low Cytotoxicity
Bitte beachten For research use only.