LysoBrite™ Blue
Lieferant: AAT BIOQUEST
22642
AATB22642EA 230 EUR
AATB22642
LysoBrite™ Blue
Tests Zelltests
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
  • Lysosome-Specific Dye
  • Long-Term Use
  • Low Cytotoxicity
Bitte beachten For research use only.