Cell Navigator® Lysosome Staining Kit *Orange Fluorescence*
Lieferant: AAT BIOQUEST
22657
AATB22657EA
230
EUR
AATB22657
Cell Navigator® Lysosome Staining Kit *Orange Fluorescence*
Tests
Zelltests
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Bitte beachten For research use only.