Tide Fluor™ 3 acid, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF3 acid
2268
AATB2268EA
307
EUR
AATB2268
Tide Fluor™ 3 acid, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3 acid
Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
559,7 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...