Tide Fluor™ 3 maleimide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF3 maleimide

2270
AATB2270EA 307 EUR
AATB2270
Tide Fluor™ 3 maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3 maleimide

Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 651,71 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5dfb4999a8243c0e380627a7db0972b89bf1a25&vid=9f462652e32049a58274812b9c32e1213b08aa84&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR