Tide Fluor™ 3 succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF3 SE
2271
AATB2271EA
307
EUR
AATB2271
Tide Fluor™ 3 succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3 succinimidyl ester
Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
555,58 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...