Tide Fluor™ 3 phosphoramidite, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF3 CEP
2274
AATB2274EA
614
EUR
AATB2274
Tide Fluor™ 3 phosphoramidite, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3 phosphoramidite
Tide Fluor™ 3 (TF3) phosphoramidite is a high-performance alternative to Cy3 phosphoramidite for the in-synthesis labeling of oligonucleotides.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
1045,36 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...