Tide Fluor™ 5WS azide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF5WS azide

2275
AATB2275EA 307 EUR
AATB2275
Tide Fluor™ 5WS azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 5WS azide

Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 933 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95d7245b12d647693abd772aa228e71f3a487717&vid=a55da5b525054d080cda66b3457f277e1c1d4422&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR