Cell Navigator® NBD Ceramide Golgi Staining Kit *Green Fluorescence*
Lieferant: AAT BIOQUEST
22750
AATB22750EA 307 EUR
AATB22750
Cell Navigator® NBD Ceramide Golgi Staining Kit *Green Fluorescence*
Tests Zelltests
The Golgi apparatus is a complex of vesicles and folded membranes within the cytoplasm of most eukaryotic cells, involved in secretion and intracellular transport.
  • live cell labeling
  • Improved signal
  • Multiple research applications
Bitte beachten For research use only.