Tide Fluor™ 5WS alkyne, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF5WS alkyne

2276
AATB2276EA 307 EUR
AATB2276
Tide Fluor™ 5WS alkyne, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 5WS alkyne

Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 787,96 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62b0c0820fa0200a3143392d87ef019a82d3ee72&vid=5122e794662e298d5a28538b1d875eae33984e3e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR