Tide Fluor™ 5WS acid, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF5WS acid

2278
AATB2278EA 460 EUR
AATB2278
Tide Fluor™ 5WS acid, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 5WS acid

Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 864,92 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb4dbac1d1bbabfb5bad0c881035c2cbf36602a5&vid=335210e18a51a4af29e4881abbcc325e8c23212a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR