Tide Fluor™ 5WS amine, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF5WS amine
2279
AATB2279EA
307
EUR
AATB2279
Tide Fluor™ 5WS amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 5WS amine
Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
1021,03 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...