Tide Fluor™ 5WS maleimide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF5WS maleimide

2280
AATB2280EA 307 EUR
AATB2280
Tide Fluor™ 5WS maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 5WS maleimide

Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 987,05 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=600591ef79bf76fbdf8cabff4ec3e04a5e2ffe53&vid=253c7f20987682134609646765ce268900ed499f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR