Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*
Lieferant: AAT BIOQUEST
22807
AATB22807EA 307 EUR
AATB22807
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*
Tests Zelltests
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for microplate readers but suitable for all major fluorescence platforms
Bitte beachten For research use only.