Cell Meter™ Caspase 3/7 Activity Apoptosis Assay Kit *Green Fluorescence Optimized for Flow Cytometry*
Lieferant: AAT Bioquest
22823
AATB22823EA 307 EUR
AATB22823
Cell Meter™ Caspase 3/7 Activity Apoptosis Assay Kit *Green Fluorescence Optimized for Flow Cytometry*
Tests Zelltests
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimized for flow cytometry but suitable for all major fluorescence platforms
Bitte beachten For research use only.