Tide Fluor™ 4 acid, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF4 acid

2285
AATB2285EA 307 EUR
AATB2285
Tide Fluor™ 4 acid, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 4 acid

TF4 is designed to be a superior fluorophore alternative to ROX and Texas Red®.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 562,65 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27a894c7db5021f88d1680ec29064e7a551a5b02&vid=49ecc7f0f6b27a9ac560fac509a39b57502f4126&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR