Tide Fluor™ 4 amine, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF4 amine
2286
AATB2286EA
307
EUR
AATB2286
Tide Fluor™ 4 amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 4 amine
TF4 is designed to be a superior fluorophore alternative to ROX and Texas Red®.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
946,81 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...