Tide Fluor™ 4, succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF4 SE
2289
AATB2289EA
307
EUR
AATB2289
Tide Fluor™ 4, succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 4, succinimidyl ester
TF4 is designed to be a superior fluorophore alternative to ROX and Texas Red®.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
659,73 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...