Tide Fluor™ 6WS amine, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF6WS amine

2292
AATB2292EA 307 EUR
AATB2292
Tide Fluor™ 6WS amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 6WS amine

Tide Fluor™ 6WS (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1187,18 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79c5c9448c930114a6a79db22f0955dd0422b764&vid=0d05005d2b16452a2944ac65330eae8f523674db&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR