Tide Fluor™ 6WS maleimide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF6WS maleimide

2293
AATB2293EA 307 EUR
AATB2293
Tide Fluor™ 6WS maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 6WS maleimide

Tide Fluor™ 6WS (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1153,2 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9273017c558817e0eafeea288aff63e8f5bb2748&vid=73e108c9b9d2e58891f9651326ff4153c3d8d647&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR