Tide Fluor™ 6WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF6WS SE

2294
AATB2294EA 152 EUR
AATB2294
Tide Fluor™ 6WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 6WS succinimidyl ester

Tide Fluor™ 6WS (TF6) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1317,7 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a07e92547a3b21b1b9bbfd655dafbb537bc0183&vid=aff56506a069c6ee17b6e2854ce671e80151cbb3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR