Tide Fluor™ 4 azide, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF4 azide
2300
AATB2300EA
307
EUR
AATB2300
Tide Fluor™ 4 azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 4 azide
TF4 is designed to be a superior fluorophore alternative to ROX and Texas Red®.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
630,74 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...