Tide Fluor™ 6WS azide, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF6WS azide
2302
AATB2302EA
307
EUR
AATB2302
Tide Fluor™ 6WS azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 6WS azide
Tide Fluor™ 6WS (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
1193,25 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...