Tide Fluor™ 6WS alkyne, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT Bioquest
|
Achtung
|
2303
AATB2303EA
307
EUR
AATB2303
Tide Fluor™ 6WS alkyne, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 6WS alkyne
Tide Fluor™ 6 (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
ADR:
5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...