Tide Fluor™ 7WS azide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF7WS azide

2304
AATB2304EA 307 EUR
AATB2304
Tide Fluor™ 7WS azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 7WS azide

Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 959,04 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dcefd97860e4ca1fe7a2841ad15abffd1e7f83a&vid=7b88896cb9adc34467a68b8920cbeba855c8b0c3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR