Tide Fluor™ 7WS alkyne, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT Bioquest
Achtung

2305
AATB2305EA 307 EUR
AATB2305
Tide Fluor™ 7WS alkyne, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 7WS alkyne

Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19bc0367cf2b3ff0e8d8102be330fe8e4847328a&vid=49d95f45a7e38001356ca0c31aed31730677b54d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR