BSB [(trans,trans)-1-Bromo-2,5-bis-(3-hydroxycarbonyl-4-hydroxy)styrylbenzene]
Lieferant: AAT BIOQUEST
23050
AATB23050EA
307
EUR
AATB23050
BSB [(trans,trans)-1-Bromo-2,5-bis-(3-hydroxycarbonyl-4-hydroxy)styrylbenzene]
BSB [(trans,trans)-1-Bromo-2,5-bis-(3-hydroxycarbonyl-4-hydroxy)styrylbenzene]
BSB is a cell-permeable fluorescent probe that specifically binds to and labels intracellular beta-amyloid aggregates both in vitro (Ki = 400 nM) and in vivo.
- Cell permeable
- Well-cited probe
- Multiple applications
Bitte beachten For research use only.
Formel:
C₂₄H₁₇BrO₆ Molecular Weight: 481,292 g/mol |
CAS-Nummer:
56776-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...