4-(1,3-Benzothiazol-2-yl)-N-methylaniline (BTA-1) ≥95%

Lieferant: AAT BIOQUEST

Synonyme: 2-(4'-(methylamino)phenyl)benzothiazole

23051
AATB23051EA 120 EUR
AATB23051
4-(1,3-Benzothiazol-2-yl)-N-methylaniline (BTA-1) ≥95%
4-(1,3-Benzothiazol-2-yl)-N-methylaniline (BTA-1)

BTA-1 is a fluorescent Thioflavin-T derivative that exhibits high affinity for amyloid deposits (Ki = 11 nM for Ab 40).


  • High Target Affinity
  • Bright Fluorescence


Bitte beachten For research use only.

Formel: C₁₄H₁₂N₂S
Molecular Weight: 240,33 g/mol
MDL: MFCD04974146
CAS-Nummer: 439858-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR