2-(4'-(Dimethylamino)phenyl)-6-methyl-benzothiazole (BTA-2) ≥95%
Lieferant: AAT BIOQUEST
23052
AATB23052EA 120 EUR
AATB23052
2-(4'-(Dimethylamino)phenyl)-6-methyl-benzothiazole (BTA-2) ≥95%
2-(4'-(Dimethylamino)phenyl)-6-methyl-benzothiazole (BTA-2)
BTA-2 is an uncharged derivative of Thioflavin-T that exhibits high affinity for amyloid deposits [Ki = 143 nm for Ab(1-40)] and can cross the blood brain barrier.
  • High Target Affinity
  • Bright Fluorescence
Bitte beachten For research use only.
CAS-Nummer: 10205-62-6