Thioflavin S ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
23059
AATB23059EA
120
EUR
AATB23059
Thioflavin S ≥95%
Thioflavin S
Thioflavin S is a mixture of compounds that results from the methylation of dehydrothiotoluidine with sulphonic acid.
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00071453 CAS-Nummer: 1326-12-1 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...