Tide Fluor™ 8WS azide, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF8WS azide
2306
AATB2306EA
307
EUR
AATB2306
Tide Fluor™ 8WS azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 8WS azide
Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
1125,19 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...