Tide Fluor™ 8WS alkyne, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF8WS alkyne

2307
AATB2307EA 307 EUR
AATB2307
Tide Fluor™ 8WS alkyne, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 8WS alkyne

Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1094,18 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97f74dc476a6ee0195c89b8cb549cc920a5e5111&vid=325b4e7ed9fe6a6d18934771e2933ad3798de69a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR