Congo red maleimide
Lieferant: AAT BIOQUEST
23070
AATB23070EA
402
EUR
AATB23070
Congo red maleimide
Congo red maleimide
Congo Red has been used as a common qualitative staining method for the identification of amyloids in vitro and in tissue sections.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only