Native Amanita phalloides Phalloidin (from Eukaryotic cells), AMCA (7-amino-4-methylcoumarin-3-acetic acid)
Lieferant: AAT BIOQUEST
23100
AATB23100EA
230
EUR
AATB23100
Native Amanita phalloides Phalloidin (from Eukaryotic cells), AMCA (7-amino-4-methylcoumarin-3-acetic acid)
Proteine und Peptide
This blue fluorescent phalloidin conjugate (equivalent to coumarin-labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Bitte beachten For research use only.