Native Amanita phalloides Phalloidin (from Eukaryotic cells), Fluorescein
Lieferant: AAT BIOQUEST
23101
AATB23101EA
230
EUR
AATB23101
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Fluorescein
Proteine und Peptide
This green fluorescent phalloidin conjugate (equivalent to FITC labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Bitte beachten For research use only.