Native Amanita phalloides Phalloidin (from Eukaryotic cells), TMR (Tetramethylrhodamine)
Lieferant: AAT BIOQUEST
23102
AATB23102EA 230 EUR
AATB23102
Native Amanita phalloides Phalloidin (from Eukaryotic cells), TMR (Tetramethylrhodamine)
Proteine und Peptide
This orange fluorescent phalloidin conjugate (equivalent to TRITC-labeled phalloidin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Bitte beachten For research use only.