Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 405
Lieferant: AAT BIOQUEST
23111
AATB23111EA
230
EUR
AATB23111
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 405
Proteine und Peptide
This blue fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 405-labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Bitte beachten For research use only.