Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 488
Lieferant: AAT BIOQUEST
23115
AATB23115EA
230
EUR
AATB23115
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 488
Proteine und Peptide
This green fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 488-labeled phalloidin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Bitte beachten For research use only.