Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor™ 514
Lieferant: AAT BIOQUEST
23116
AATB23116EA
230
EUR
AATB23116
Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor™ 514
Proteine und Peptide
This yellow fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 514-labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Bitte beachten For research use only.