Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 555
Lieferant: AAT BIOQUEST
23119
AATB23119EA
230
EUR
AATB23119
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 555
Proteine und Peptide
This orange fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 555-labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Bitte beachten For research use only.