Native Amanita phalloides Phalloidin (from Amanita phalloides), Alexa Fluor® 790
Lieferant: AAT BIOQUEST
23131
AATB23131EA 460 EUR
AATB23131
Native Amanita phalloides Phalloidin (from Amanita phalloides), Alexa Fluor® 790
Proteine und Peptide
This infrared fluorescent phalloidin conjugate selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
  • Brighter signal
  • High affinity and selectivity
  • In house quality control
Bitte beachten For research use only.