Phalloidin, XFD350
Lieferant: AAT BIOQUEST
23150
AATB23150EA
460
EUR
AATB23150
Phalloidin, XFD350
Proteine und Peptide
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
- Well-cited conjugate
- High affinity and selectivity
- In house quality control
Bitte beachten For research use only.